Learn More
Medchemexpress LLC P-tyramine-d4 hydrochloride | 1189884-47-6 | MFCD08704875 | 99.8% | 177.66 g·mol⁻¹ | C8H8D4ClNO | 10 MG
SDP
Catalog No. 5000300503
Encompass_Preferred
p-Tyramine-d4 hydrochloride is the deuterium-labeled hydrochloride salt of p-tyramine supplied as a solid stable isotope standard for analytical and bioanalytical workflows. It is used as an internal standard or reference in LC-MS and GC-MS quantification and method development for accurate determination of tyramine in biological and food matrices.
  • Deuterium-labeled internal standard for accurate quantification.
  • High listed purity suitable for analytical assays.
  • White to off-white solid for straightforward handling and storage.
  • Available in small pack sizes for standard preparation.
  • Molecular formula C8H8D4ClNO and molecular weight 177.66 g·mol⁻¹ aid method documentation.
Catalog No. 50-003-00503 Supplier Medchemexpress LLC Supplier No. HYW016823S10MG
p-Tyramine-d4 hydrochloride is the deuterium-labeled hydrochloride salt of p-tyramine supplied as a solid stable isotope standard for analytical and bioanalytical workflows. It is used as an internal standard or reference in LC-MS and GC-MS quantification and method development for accurate determination of tyramine in biological and food matrices.
  • Deuterium-labeled internal standard for accurate quantification.
  • High listed purity suitable for analytical assays.
  • White to off-white solid for straightforward handling and storage.
  • Available in small pack sizes for standard preparation.
  • Molecular formula C8H8D4ClNO and molecular weight 177.66 g·mol⁻¹ aid method documentation.