Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylene, Analytical Standard, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019942 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9942 5 mL
11-101-8188 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-9942 Supplier MilliporeSigma Supelco Supplier No. 956805ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-Xylene is an aromatic liquid, which is an isomeric form of xylene. It can be produced via Diels-Alder reaction between 2,5-dimethylfuran and ethylene catalyzed by zeolite.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 106-42-3
Vapor Pressure 9 mmHg (20°C)
Vapor Density 3.7 (vs. air)
Density 0.861 g/mL (at 20°C)
Physical Form Neat
Linear Formula C6H4(CH3)2
MDL Number MFCD00008556
Melting Point 12°C to 13°C
Boiling Point 138°C
Synonym 1,4-Dimethylbenzene
Quantity 5 mL
Formula Weight 106.17
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.