Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC P-xylene-bis(N-pyridinium bromide) | 14208-10-7 | MFCD00043749 | >95.0% | 422.16 g·mol⁻¹ | C18H18Br2N2 | 5 G
SDP

Catalog No. 5000283673
Encompass_Preferred

p-Xylene bis(pyridinium bromide) is a bisquaternary pyridinium salt used as a cationic charged quencher and a weak acetylcholinesterase and butyrylcholinesterase inhibitor. It is provided as a solid powder and is used in biochemical assays, staining, and histology workflows.

  • Used as a cationic charged quencher in fluorescence and staining applications.
  • Weak AChE inhibitor (IC50 ≈ 1,540 μM) and BChE inhibitor (IC50 ≈ 529 μM).
  • White to off-white solid, soluble in DMSO and DMF.
  • Purity ≥95% (TLC).
  • Molecular weight 422.16 g·mol⁻¹; formula C18H18Br2N2.

Catalog No. 50-002-83673 Supplier Medchemexpress LLC Supplier No. HYW1300255G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

p-Xylene bis(pyridinium bromide) is a bisquaternary pyridinium salt used as a cationic charged quencher and a weak acetylcholinesterase and butyrylcholinesterase inhibitor. It is provided as a solid powder and is used in biochemical assays, staining, and histology workflows.

  • Used as a cationic charged quencher in fluorescence and staining applications.
  • Weak AChE inhibitor (IC50 ≈ 1,540 μM) and BChE inhibitor (IC50 ≈ 529 μM).
  • White to off-white solid, soluble in DMSO and DMF.
  • Purity ≥95% (TLC).
  • Molecular weight 422.16 g·mol⁻¹; formula C18H18Br2N2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.