Learn More
Medchemexpress LLC p-xylene bis(pyridinium bromide) | 14208-10-7 | MFCD00043749 | 99.6% | 422.16 g/mol | C18H18Br2N2 | 1 G
SDP
Catalog No. 5000282582
Encompass_Preferred
p-Xylene bis(pyridinium bromide) is a bisquaternary pyridinium compound used in biochemical research as a cationic charged quencher and a weak inhibitor of acetylcholinesterase and butyrylcholinesterase. Its reported activity and high purity make it suitable for cholinesterase inhibitor studies and related biochemical assays.
  • Cationic charged quencher for fluorescence assays.
  • Weak bisquaternary AChE inhibitor (IC50 = 1540 μM).
  • Weak bisquaternary BChE inhibitor (IC50 = 529 μM).
  • High purity suitable for research use (99.6%).
  • Molecular weight 422.16 g/mol; formula C18H18Br2N2.
  • Supplied as a 1 g pack for laboratory use.
Catalog No. 50-002-82582 Supplier Medchemexpress LLC Supplier No. HYW1300251G
p-Xylene bis(pyridinium bromide) is a bisquaternary pyridinium compound used in biochemical research as a cationic charged quencher and a weak inhibitor of acetylcholinesterase and butyrylcholinesterase. Its reported activity and high purity make it suitable for cholinesterase inhibitor studies and related biochemical assays.
  • Cationic charged quencher for fluorescence assays.
  • Weak bisquaternary AChE inhibitor (IC50 = 1540 μM).
  • Weak bisquaternary BChE inhibitor (IC50 = 529 μM).
  • High purity suitable for research use (99.6%).
  • Molecular weight 422.16 g/mol; formula C18H18Br2N2.
  • Supplied as a 1 g pack for laboratory use.