Learn More
Medchemexpress LLC P-xylene bis(pyridinium bromide) | 14208-10-7 | MFCD00043749 | 99.6% | 422.16 g/mol | C18H18Br2N2 | 100 MG
SDP
Catalog No. 5000282732
Encompass_Preferred
p-Xylene bis(pyridinium bromide) is a cationic charged quencher supplied as a high-purity solid for research use. It acts as a weak bisquaternary inhibitor of acetylcholinesterase and butyrylcholinesterase and is used in fluorescence quenching, membrane fusion, and biochemical assay applications.
  • High-purity solid suitable for research (99.6%).
  • Cationic charged quencher used in fluorescence and FRET assays.
  • Weak bisquaternary inhibitor of AChE and BChE (hAChE IC50 1540 μM; BChE IC50 529 μM).
  • Molecular formula C18H18Br2N2; molecular weight 422.16 g/mol.
  • Supplied in 100 mg packaging; store sealed away from moisture at 4°C (in solvent: -20°C for 1 month; -80°C for 6 months).
Catalog No. 50-002-82732 Supplier Medchemexpress LLC Supplier No. HYW130025100MG
p-Xylene bis(pyridinium bromide) is a cationic charged quencher supplied as a high-purity solid for research use. It acts as a weak bisquaternary inhibitor of acetylcholinesterase and butyrylcholinesterase and is used in fluorescence quenching, membrane fusion, and biochemical assay applications.
  • High-purity solid suitable for research (99.6%).
  • Cationic charged quencher used in fluorescence and FRET assays.
  • Weak bisquaternary inhibitor of AChE and BChE (hAChE IC50 1540 μM; BChE IC50 529 μM).
  • Molecular formula C18H18Br2N2; molecular weight 422.16 g/mol.
  • Supplied in 100 mg packaging; store sealed away from moisture at 4°C (in solvent: -20°C for 1 month; -80°C for 6 months).