Learn More
Medchemexpress LLC p-xylene bis(pyridinium bromide) | 14208-10-7 | MFCD00043749 | 99.6% | 422.16 g·mol⁻¹ | C18H18Br2N2 | 250 MG
SDP
Catalog No. 5000282940
Encompass_Preferred
p-Xylene bis(pyridinium bromide) is a bisquaternary pyridinium compound provided as a research reagent and cationic charged quencher. It is reported as a weak inhibitor of acetylcholinesterase and butyrylcholinesterase (IC50 hAChE 1540 μM; BChE 529 μM), and is suitable for biochemical assays and probe development.
  • High purity: 99.6%.
  • Useful as a cationic charged quencher in fluorescence and biochemical assays.
  • Characterized inhibitory activity against AChE and BChE (IC50 hAChE 1540 μM; BChE 529 μM).
  • Supplied as a solid 250 mg pack for laboratory use.
  • Molecular formula C18H18Br2N2 and molecular weight 422.16 g·mol⁻¹.
Catalog No. 50-002-82940 Supplier Medchemexpress LLC Supplier No. HYW130025250MG
p-Xylene bis(pyridinium bromide) is a bisquaternary pyridinium compound provided as a research reagent and cationic charged quencher. It is reported as a weak inhibitor of acetylcholinesterase and butyrylcholinesterase (IC50 hAChE 1540 μM; BChE 529 μM), and is suitable for biochemical assays and probe development.
  • High purity: 99.6%.
  • Useful as a cationic charged quencher in fluorescence and biochemical assays.
  • Characterized inhibitory activity against AChE and BChE (IC50 hAChE 1540 μM; BChE 529 μM).
  • Supplied as a solid 250 mg pack for laboratory use.
  • Molecular formula C18H18Br2N2 and molecular weight 422.16 g·mol⁻¹.