Learn More
Sigma Aldrich Fine Chemicals Biosciences p Xylene ReagentPlusR 99
Catalog No. 502898072
Encompass
p-Xylene is an isomeric form of xylene. It undergoes friedel-Crafts acylation reaction with various acids in the presence of zeolites (catalyst). It can be obtained from the mixtures of isomeric forms of xylene and ethylbenzene isolated from the naptha fraction of petroleum. Also it can be prepared from toluene by employing ZSM (Zeolite Socony Mobil)-5 catalysts. It is a raw material for the manufacture of PET (poly(ethylene terephthalate)) bottles. Its sustainable synthesis from ethylene and dimethylfuran in the presence of zeolite catalysts has been reported.
Catalog No. 50-289-8072 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1344494L
p-Xylene is an isomeric form of xylene. It undergoes friedel-Crafts acylation reaction with various acids in the presence of zeolites (catalyst). It can be obtained from the mixtures of isomeric forms of xylene and ethylbenzene isolated from the naptha fraction of petroleum. Also it can be prepared from toluene by employing ZSM (Zeolite Socony Mobil)-5 catalysts. It is a raw material for the manufacture of PET (poly(ethylene terephthalate)) bottles. Its sustainable synthesis from ethylene and dimethylfuran in the presence of zeolite catalysts has been reported.