Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylenol Blue 95.0+%, TCI America™
SDP

Catalog No. X001625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
X001625G 25 g
X00161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. X001625G Supplier TCI America Supplier No. X001625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 125-31-5
Molecular Formula C23H22O5S
Molecular Weight (g/mol) 410.48
MDL Number MFCD00005870
InChI Key MGUKYHHAGPFJMC-UHFFFAOYSA-N
PubChem CID 67172
IUPAC Name 3,3-bis(4-hydroxy-2,5-dimethylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
SMILES CC1=CC(=C(C)C=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C=C1C

Specifications

Color Purple/Red
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 410.48
Percent Purity ≥95.0% (HPLC)
Chemical Name or Material p-Xylenol Blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.