Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xyloquinone 98.0+%, TCI America™
SDP

Catalog No. D06861G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D06861G 1 g
D06865G 5 g
D068625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D06861G Supplier TCI America Supplier No. D06861G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 137-18-8
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00041737
InChI Key MYKLQMNSFPAPLZ-UHFFFAOYSA-N
Synonym 2,5-dimethyl-1,4-benzoquinone, phlorone, p-xyloquinone, 2,5-dimethyl-p-benzoquinone, 2,5-xyloquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-dimethyl, 2,5-dimethylquinone, floron, 3,6-dimethyl-p-benzoquinone, floron czech
PubChem CID 8718
IUPAC Name 2,5-dimethylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C(=CC1=O)C

Specifications

Melting Point 125°C
Quantity 1 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material p-Xyloquinone
RTECSNumber DK4800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.