Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

p-Xylylenebis(triphenylphosphonium bromide), 96%

Catalog No. AAA1824122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1824122 100 g
AAA1824114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1824122 Supplier Thermo Scientific Chemicals Supplier No. A1824122

missing translation for 'validMainframeMsg'

CAS: 40817-03-6 | C44H38Br2P2 | 788.54 g/mol

p-Xylylenebis(triphenylphosphonium bromide) is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
p-Xylylenebis(triphenylphosphonium bromide) is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40817-03-6
Molecular Formula C44H38Br2P2
Molecular Weight (g/mol) 788.54
MDL Number MFCD00011914
InChI Key ZZQVVCXWFPGKJD-UHFFFAOYSA-L
Synonym 1,4-phenylenebis methylene bis triphenylphosphonium bromide, p-xylylenebis triphenylphosphonium bromide, 1,4-phenylenebis methylene bis triphenylphosphonium dibromide, rarechem fh 1w 0055, phosphonium,1,1'-1,4-phenylenebis methylene bis 1,1,1-triphenyl-, bromide 1:2, triphenyl 4-triphenylphosphaniumyl methyl phenyl methyl phosphanium dibromide, triphenyl-4-triphenylphosphaniumylmethyl phenyl methyl phosphanium dibromide
PubChem CID 2723797
SMILES [Br-].[Br-].C(C1=CC=C(C[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 3584592
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water.
IUPAC Name triphenyl({4-[(triphenylphosphaniumyl)methyl]phenyl}methyl)phosphanium dibromide
Formula Weight 788.55
Percent Purity 96%
Chemical Name or Material p-Xylylenebis(triphenylphosphonium bromide)
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 255-092-7
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.