Learn More
ABclonal Technology p-YWHAZ/H/G/B-S58 /S59/S59/S60
SDP
Supplier:  ABclonal Technology AP1561100UL
Encompass_Preferred
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins This highly conserved protein family is found in both plants and mammals and this protein is 99% identical to the mouse rat and sheep orthologs The encoded protein interacts with IRS1 protein suggesting a role in regulating insulin sensitivity Several transcript variants that differ in the 5 UTR but that encode the same protein have been identified for this gene
Catalog No. 50-000-13717