Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ 384-Well Clear Polystyrene Microplates
GREENER_CHOICE

Catalog No. 07201101 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-201-101 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-201-101 Supplier Corning™ Supplier No. 3640
Add to Cart
Edge
Add to Cart

Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ 364-Well Clear Polystyrene Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 364-well microplates for laboratory miniaturization and automation.

  • Total well volume of 112 μL; working well volume of 20 μL – 80 μL
  • Cell culture microplates are sterile and nonpyrogenic.
  • The 384-well Universal Optics NBS microplate is manufactured using an advanced polymer with high clarity and improved chemical resistant properties.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid Without Lid
    Description Nonbinding (NBS™) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions
    Array 16 x 24
    Color Clear
    Product Type 384-Well NBS™ Microplate
    Material Polystyrene
    No. of Wells 384
    Volume (Metric) Working 20 to 80 μL
    Sterility Non-sterile
    Surface Treatment Non-treated
    Total Volume (Metric) Well 112 μL
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Reducing protein and nucleic acid binding at low concentrations and increasing assay signal to noise
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Well Color Clear
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.