Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Sharps-A-Gator™ Sharps Containers

Catalog No. 14-827-63 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (English):
1 gal.
2 gal.
7 gal.
7.5 gal.
10 gal.
Capacity (Metric):
3.8 L
7.5 L
26.5 L
28 L
38 L
Closure Type:
Lockable
Slide Top
Shape:
Rectangle
Round
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Capacity (Metric) Closure Type Shape
14-827-63 1 gal. 3.8 L Lockable Rectangle
14-827-104 7 gal. 26.5 L Lockable Rectangle
14-827-105 7.5 gal. 28 L Lockable Round
14-827-71 10 gal. 38 L Lockable Rectangle
22-037-959 2 gal. 7.5 L Slide Top Rectangle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 14-827-63
This item is not returnable. View return policy

Puncture-resistant with wide rectangular opening for horizontal loading of needles

  • General-purpose sharps container with clear locking lid and marked with biohazard label
  • Made from puncture-resistant polypropylene that can be autoclaved or incinerated
  • Easily mount on walls and carts, nest to conserve storage space

Compliance

Used to comply with OSHA standards for sharps and biohazard waste collection.

Specifications

Capacity (Metric) 3.8 L
Certifications/Compliance Used to comply with OSHA standards for sharps and biohazard waste collection.
Closure Type Lockable
Closure Color Clear
Capacity (English) 1 gal.
Color Red
Dimensions (L x W x H) 7 x 10.5 x 6.25 in.
Disposable Single-use
Height (Metric) 16 cm
Length (Metric) 18 cm
Quantity 32/Cs.
Shape Rectangle
Width (Metric) 27 cm
Autoclavable Autoclavable
Width (English) 10.5 in.
Length (English) 7 in.
Height (English) 6.25 in.
Material Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.