Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Disposable Cuvettes

Catalog No. 14955125 Shop All Fisherbrand Products
Change view
Click to view available options
Capacity (Metric):
1.5 mL
4.5 mL
Type:
Cuvette
Semi-micro Cell
Standard Cuvette
Material:
Methacrylate
PMMA
Polystyrene
Wavelength Range:
285 to 750 nm
340 to 750 nm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Type Material Wavelength Range
14-955-125 4.5 mL Standard Cuvette Polystyrene 340 to 750 nm
14-955-129 4.5 mL Cuvette Polystyrene 340 to 750 nm
14-955-126 4.5 mL Standard Cuvette PMMA 285 to 750 nm
14-955-130 4.5 mL Cuvette PMMA 285 to 750 nm
14-955-127 1.5 mL Semi-micro Cell Polystyrene 340 to 750 nm
14-955-128 1.5 mL Semi-micro Cell Methacrylate 285 to 750 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-955-125
Only null left
Add to Cart
Add to Cart

Choice of polystyrene or methacrylate

Fisherbrand™ Disposable Cuvettes are available in choice of polystyrene or methacrylate. Select polystyrene cuvets for assays in visible spectral range (340 to 750nm); for accuracy throughout UV/Vis range (285 to 750nm), choose cuvets made from a superior grade of methacrylate.

  • Disposable 10mm pathlength cuvets designed for visible and UV/Vis assays
  • Clear four-sided cuvets are ideal for fluorimetry and nephelometry, as well as spectrophotometry
  • Scale down assay volumes and save on reagents by using semimicro cuvets with capacity to 1.5mL
  • Molded-in arrow indicates direction of transmission on standard and semimicro cuvets—helping assure uniformity
  • Each case contains cuvets with same mold impression number—assuring precision between samples—standard deviation is <±0.005 extinction units between cuvets

Order Info

Packaged 100 cuvets per stackable tray; five trays to a case. Outside dimensions: 0.47L x 0.47W x 1.77 in. H (12 x 12 x 45mm).

Specifications

Type Standard Cuvette
Disposable Single-use
Capacity (Metric) 4.5 mL
Material Polystyrene
Length (Metric) 12mm
Quantity 500 Case
Path Length 10 mm
Wavelength Range 340 to 750 nm
Height (Metric) 45 mm
For Use With (Equipment) Spectrophotometers, Fluorometers, Nephelometers
Width (Metric) 12 mm
For Use With (Application) For assays in visible spectral range, accuracy throughout UV/Visible range
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.