Learn More
Fisherbrand™ Chemical Splash and Impact Goggles
Catalog No. 19-181-504 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Chemical Splash Protection Goggles
Impact Goggles
Ventilation Details:
Four indirect vents
Perforations on goggle top and sides provide direct ventilation
Certifications/Compliance:
ANSI/ISEA Z87.1-2020: Z87+ U6
ANSI/ISEA Z87.1-2020: Z87+ U6 D3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Ventilation Details Certifications/Compliance
19-181-504 Chemical Splash Protection Goggles Four indirect vents ANSI/ISEA Z87.1-2020: Z87+ U6 D3
19-181-503 Impact Goggles Perforations on goggle top and sides provide direct ventilation ANSI/ISEA Z87.1-2020: Z87+ U6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-181-504
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lens coated to minimize scratching and fogging.
Lens coated to minimize scratching and fogging
  • Built for comfortable protection with a soft, flexible PVC body that conforms to facial contours
  • Large frames fit over most prescription eyewear
  • Coated, clear polycarbonate lens resists impact, scratching, and fogging
  • Headstrap easily adjusts for a custom fit
  • Resist a wide range of chemicals and feature indirect ventilation to direct airflow over the lens
Order Info
NOTE: 19-181-503 is the IMPACT style goggles; 19-181-504 are the CHEMICAL SPLASH style goggles.
Compliance
Meets ANSI Z87.1 and ANSI Z80.3 standards.
Specifications
Certifications/Compliance ANSI/ISEA Z87.1-2020: Z87+ U6 D3
Lens Coating Anti-fog, Scratch-resistant
Shape Round
Lens Tint Clear
Product Type Chemical Splash Protection Goggles
Frame Color Green
Ventilation Details Four indirect vents
Material Polycarbonate
Lens Material Polycarbonate
Frame Material PVC