Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ L-Glutamine (200 mM)
GREENER_CHOICE

Catalog No. 25030081
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL
100 mL
20 x 100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-030-081 100 mL
25-030-149 20 mL
25-030-164 20 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 25-030-081 Supplier Gibco™ Supplier No. 25030081
Add to Cart
Add to Cart
This item is not returnable. View return policy

Gibco L-Glutamine is a ready-to-use amino acid supplement for cell culture. L-glutamine is an amino acid required in cell culture for a variety of critical cellular processes.

Gibco L-Glutamine is a ready-to-use amino acid supplement for cell culture. L-glutamine is an amino acid required in cell culture for the formation of purine and pyrimidine nucleotides, amino sugars, glutathione, L-glutamate, and other amino acids, as well as in protein synthesis and glucose production. Unlike most other amino acids, L-glutamine is not stable in solution with a rate of degradation dependent upon time, temperature, and pH. The optimal concentration depends upon the cell type and medium used to culture the cells, but generally falls in the range of 2–6 mM.

Gibco L-Glutamine supplement is a ready-to-use 200 mM stock solution for cell culture supplementation that supports of many critical cellular functions, including:

  • Formation of purine and pyrimidine nucleotides
  • Formation of amino sugars, glutathione, and L-glutamate
  • Formation of other amino acids
  • Protein synthesis
  • Glucose production

cGMP manufacturing and quality system

For supply chain continuity, we manufacture this product at two separate facilities located in Grand Island, NY and Scotland, UK. Both sites are compliant with cGMP manufacturing requirements and are certified to the ISO 13485 standard.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glutamine
CAS 56-85-9
pH 5 to 6
Packaging Bottle
Molecular Weight (g/mol) 146.14 g/mol
Quantity 100 mL
Grade Cell Culture
Concentration or Composition (by Analyte or Components) 100X
Physical Form Liquid
Product Line Gibco
Shipping Condition Dry Ice
Show More Show Less
Why did my 200 mM L-Glutamine precipitate out of solution when I thawed it?

When L-glutamine is in a concentrated stock solution it easily precipitates when cooled. Warming the solution briefly in a 37C water bath with gentle swirling will dissolve the precipitate. Do not use the product unless the precipitate is fully dissolved.

Why does L-Glutamine (200 mM) (Cat. No. 25050149, 25030081, 25030164) solution have “Product of Brazil” on the label?

L-Glutamine (200 mM) (Cat. No. 25050149, 25030081, 25030164) solution is made in the USA from a raw material glutamine powder manufactured in Brazil. The country of manufacture of a component that has been subjected to manufacturing or processing operations in more than one country is the last country in which the component has been “substantially transformed”. A “substantial transformation” occurs in a country when a product emerges from a manufacturing process with a name, character, or use different from that of the original material(s) subjected to that process.

Diluting, purifying, sorting, testing, packing, repacking, or affixing labels to a product in a country does not constitute a “substantial transformation” in that country, hence the label must bear the country of the original raw material origin.

My order of L-Glutamine (Cat. No. 25030081) arrived frozen. Is it stable?

If L-Glutamine is completely or partially frozen, it is still stable. It becomes unstable when stored for extended periods of time completely thawed, especially above 2-8 degrees C.

What is your recommended method for thawing L-Glutamine before adding to cell culture media?

We recommend thawing at 2-8 degrees C and then warming at 37 degrees C until the material goes into solution. Mix it as it warms up to spend minimal time at elevated temperatures before you aliquot it.

Why did my 200 mM L-glutamine precipitate when I thawed it?

When L-glutamine is in a concentrated stock solution it easily precipitates when cooled. Warming the solution briefly in a 37 degrees C water bath with gentle swirling will dissolve the precipitate. Do not use the product unless the precipitate is fully dissolved.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.