Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nitric Acid (Certified ACS Plus), Fisher Chemical™
GSA_VA

Catalog No. A200500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
6.5 gal.
4 x 2.5 L
Packaging:
Glass Bottle
Glass Carboy
Safe-Cote™ Glass
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A200-500 500 mL Glass Bottle
A200S-500 500 mL Safe-Cote™ Glass
A200-212 2.5 L Glass Bottle
A200S-212 2.5 L Safe-Cote™ Glass
A200C-212 4 x 2.5 L Glass Bottle
A200-612GAL 6.5 gal. Glass Carboy
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. A200-500
  • $214.00
    $164.65 / Each

    Save $49.35

    23% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $984.50
    $766.65 / Case of 6 EA

    Save $217.85

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
missing translation for 'nonReturnableMessage' missing translation for 'returnPolicyLabel'

Nitric Acid material may darken during storage due to photochemical reaction

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7697-37-2 , 7732-18-5
Molecular Formula HNO3
Molecular Weight (g/mol) 63.01
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
Synonym hydrogen nitrate, aqua fortis, azotic acid, salpetersaeure, rfna, acidum nitricum, nital, acide nitrique, nitrous fumes, nitryl hydroxide
PubChem CID 944
ChEBI CHEBI:48107
IUPAC Name nitric acid
SMILES O[N+]([O-])=O

Specifications

pH -1.0
Color Colorless Yellow
Melting Point -41°C
Boiling Point 120.5°C
Density 1.40g/cm³
Heavy Metals (as Pb) 0.2ppm max.
Identification Pass Test
Packaging Glass Bottle
Residue after Ignition 2ppm max.
Vapor Pressure 0.94 kPa at 20°C
Normality about 15.8
Assay Percent Range 70%
Quantity 500 mL
Physical Form Liquid
UN Number 2031
Formula Weight 63.01
Percent Purity 68.0 to 70.0% (w/w)
Grade Certified ACS Plus
Chemical Name or Material Nitric Acid
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause pulmonary edema. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required.

NFPA

Health:4
Flammability:0
Instability:0
DOTInformation DOT Class 8, : Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.