Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNaseZap™ RNase Decontamination Solution

Catalog No. AM9780
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
4 L
6 x 250 mL
Product Type:
RNase
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
AM9780 250 mL RNase
AM9782 6 x 250 mL RNase
AM9784 4 L RNase
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AM9780 Supplier Invitrogen™ Supplier No. AM9780
Get 15% Off Sample Preparation Products
Use promo code SAMPREP15
View all promotions
Add to Cart
Edge
Add to Cart

RNaseZap™ RNase Decontamination Solution is a surface decontamination solution that destroys RNases on contact. You simply spray RNaseZap™ Solution onto the surface to be decontaminated and rinse it off with RNase-free water.

RNaseZap™ RNase Decontamination Solution is a surface decontamination solution that destroys RNases on contact. You simply spray RNaseZap™ Solution onto the surface to be decontaminated and rinse it off with RNase-free water.

Features of RNaseZap™ RNase Decontamination Solution:

  • Completely removes RNase contamination from glass and plastic surfaces
  • Excels at removing high levels of RNase contamination whereas similar products fail
  • Proven effective at removing high concentrations of dried-on RNase A
  • Ideal for cleaning work surfaces, pipettors, and equipment that must be RNase-free

Using RNaseZap™ Solution

Working with RNA requires that special measures be taken to ensure an RNase-free environment. Even trace quantities of RNase can lead to lower yields from in vitro transcription reactions, degradation during RNA purification protocols, and variable results in RPAs and Northerns. RNaseZap™ contains three ingredients active against RNase and has proven to be extremely effective at removing RNase contamination from glassware, plastic surfaces, countertops, and pipettors. It has also been shown to be effective at eliminating RNase contamination from microfuge tubes without inhibiting subsequent enzymatic reactions. Supplied in one 250 mL bottle.

RNase contamination in microfuge tubes

While concern about RNase contamination of frequently handled labware is common and prudent, researchers often do not worry about RNase contamination in commercially prepared products such as microfuge tubes. However, in a survey of six standard 1.5 mL microfuge tubes in which three were pre-rinsed with RNaseZap™ and water and three were left untreated, all of the untreated tubes had at least marginal levels of RNase contamination, whereas none of the tubes pre-rinsed with RNaseZap™ exhibited contamination. Additionally, in testing commercially available 'RNase-free' microfuge tubes, approximately 10% had some RNase contamination (data not shown). This may well account for the erratic results that researchers periodically experience when performing RNA-based assays.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Product Type RNase
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Quantity 250 mL
Reagent Type RNase Control Reagent
Product Line Ambion, RNaseZap
What is the formulation and pH of RNaseZAP Rnase Decontamination Solution (Cat. Nos. AM9782, AM9784, AM9780)?

The formulation for RNaseZAP RNase Decontamination Solution (Cat. Nos. AM9782, AM9784, AM9780) is proprietary. The pH is 7.0-7.4.

What is the level of detection for somatic variants with the Oncomine Myeloid Research Assay GX?

Detection of somatic variants with the Oncomine Myeloid Research Assay GX has been verified down to 5% allele frequency.

Which extraction kits are compatible with Oncomine Myeloid Research Assay GX?

Here is a list of kits that are compatible with Oncomine Myeloid Research Assay GX:

Nucleic acid isolation - RNA samples
- MagMAX mirVana Total RNA Isolation Kit, Cat. No. A27828
- PureLink Total RNA Blood Kit, Cat. No. K156001
- RNaseZap RNase Decontamination Solution, Cat. No. AM9780, AM9782, AM9784

Nucleic acid isolation - DNA samples
- MagMAX DNA Multi-Sample Ultra 2.0 Kit, Cat. No. A36570
- PureLink Genomic DNA Mini Kit, Cat. No. K1820-01

What are the sample input requirements for Oncomine Myeloid Research Assay GX?

The recommended minimum sample input is 27.75 ng of DNA (1.11 ng/µL minimum concentration) and 14.25 ng of RNA (0.95 ng/µL minimum concentration).

What sample controls should I use with the Oncomine Myeloid Research Assay GX?

The recommended controls are:

DNA Controls:
- Seraseq® Myeloid Mutation DNA Mix, Cat. No. 0710-0408 (SeraCare®)
- Myeloid DNA Reference Standard, Cat. No. HD829 (Horizon™)
- AcroMetrix™ Oncology Hotspot Control, Cat. No. 969056

RNA Controls:
- Universal Human Reference RNA, Cat. No. 740000 (Agilent™)
- Seraseq® Myeloid Fusion RNA Mix, Cat. No. 0710-0407 (SeraCare®)

How many Oncomine Myeloid Research Assay GX samples can be multiplexed on a GX5 Chip?

Multiplex sequencing of up to 8 Oncomine Myeloid Research Assay GX samples (8 DNA+ 8 RNA) per lane on a GX5 Chip can be performed in a single run. There are 4 lanes on the GX5 Chip, so 32 total samples (32 DNA+ 32 RNA) can be run on one GX5 Chip (8 samples per run).

What are the components of Oncomine Myeloid Research Assay GX?

The Oncomine Myeloid Research Assay GX (Cat. No. A47857) includes the 2-pool DNA panel, the 1-pool RNA panel, Genexus Strip 1, and Genexus Strip 2-AS. The DNA panel is provided in two packs of 8 tubes (4 tubes of Myeloid DNA Pool 1 and 4 tubes of Myeloid DNA Pool 2 per pack). The RNA panel is provided in one pack of 8 tubes (Myeloid RNA Pool 1). Each primer pool in the panel is provided in pairs of tubes, where each tube pair contains one tube with primers in position 1 and one empty uncapped tube in position 2. Three 8-strip packs of the Genexus Strip 1 and Genexus Strip 2-AS (24 total of each strip) are provided with each kit.

How many samples can I run using Oncomine Myeloid Research Assay GX?

The contents of each Oncomine Myeloid Research Assay GX kit are sufficient for up to 32 samples (32 x 3‑pool reactions: 32 x 2‑pool DNA reactions and 32 x 1‑pool RNA reactions).

How many primer pools are present in the Oncomine Myeloid Research Assay GX?

The Oncomine Myeloid Research Assay GX includes 3 pools of Ion AmpliSeq oligonucleotide primers (a 2-pool DNA panel and a 1-pool RNA panel).

What is the Oncomine Myeloid Research Assay GX gene content?

The Oncomine Myeloid Research Assay GX targets key genes and fusions associated with major myeloid disorders, including acute myeloid leukemia (AML), myeloid dysplastic syndrome (MDS), myeloproliferative neoplasms (MPN), chronic myeloid leukemia (CML), chronic myelomonocytic leukemia (CMML), and juvenile myelomonocytic leukemia (JMML). The DNA panel comprises 40 key genes while the RNA panel includes a broad fusion panel of 29 driver genes, enabling detection over 600 unique fusion isotypes. See flyer for gene targets (https://assets.thermofisher.com/TFS-Assets/CSD/Flyers/Oncomine_myeloid_assay_gx_flyer.pdf).

What is the Oncomine Myeloid Research Assay GX designed for ?

The Oncomine Myeloid Research Assay GX is a comprehensive targeted next-generation sequencing (NGS) assay designed for sensitive detection of myeloid disorder-associated DNA mutations and fusion transcripts in blood and bone marrow samples.


For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.