Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAZap™ PCR DNA Degradation Solutions

Catalog No. AM9890
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AM9890 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AM9890 Supplier Invitrogen™ Supplier No. AM9890
Get 15% Off Sample Preparation Products
Use promo code SAMPREP15
View all promotions
Add to Cart
Edge
Add to Cart

DNAZap™ Solutions are two solutions that are innocuous when used alone, but become a potent nucleic acid degrading reagent when mixed.

DNAZap™ Solutions are two solutions that are innocuous when used alone, but become a potent nucleic acid degrading reagent when mixed. This mixture is able to instantaneously degrade high levels of contaminating DNA and RNA from surfaces. Features of DNAZap™ Solutions:

  • Completely degrades contaminating DNA and RNA at the level of PCR sensitivity
  • Works on contact
  • Degrades nucleic acid, unlike other products on the market that only act as detergents
  • Ideal for cleaning PCR tubes, PCR machine surfaces, pipettors, lab benches, lab equipment, and microfuge tubes
TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
Content And Storage DNAZap™ Solution 1 and DNAZap™ Solution 2 should be stored at 2–8°C.
Shipping Condition Room Temperature
Quantity 250 mL
Reagent Type DNA Decontamination Reagent
Product Line Ambion, DNAZap
What are the compositions DNAZap PCR DNA Degradation Solutions (Cat. No. AM9890)?

The compositions of these buffers are proprietary.

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.