Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TAE Buffer, Tris-Acetate-EDTA, 50X Solution, Electrophoresis, Fisher BioReagents™
GSA_VA

Catalog No. BP13321 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
Poly CUBE
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP13321 1 L Nalgene Poly Bottle
BP1332500 500 mL Poly Bottle
BP13324 4 L Poly CUBE
BP133220 20 L Poly CUBE
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. BP13321
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Tris-Acetate-EDTA, CAS Number-77-86-1, 60-00-4, 6850-28-8, TAE, 4L, Gray, Tris (24%), Acetic Acid (5.0%), and EDTA (<2%)., DNase free, Pass Test, Filtered through a 0.2-micron filter., Electrophoresis, 50X Solution, Poly CUBE, Liquid, Protease free, DNase-, RNase- and Protease-Free, RT

Tris-Acetate-EDTA, CAS Number-77-86-1, 60-00-4, 6850-28-8, TAE, 4L, Gray, Tris (24%), Acetic Acid (5.0%), and EDTA (<2%)., DNase free, Pass Test, Filtered through a 0.2-micron filter., Electrophoresis, 50X Solution, Poly CUBE, Liquid, Protease free, DNase-, RNase- and Protease-Free, RT

  • Save time and simplify your buffer preparation step by using Fisher BioReagents 50X TAE Solution - simply dilute as needed!
  • 50X solution contains 2M tris-acetate and 0.050M EDTA
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 50X TAE
  • Solution with 49 volumes of ultrapure water, such as BP2819
  • Tested for the absence of DNase, RNase, and protease
  • Filtered, autoclaved, and functionally tested for use in electrophoresis applications
  • TAE is a commonly used buffer for electrophoresis of nucleic acids in agarose gels
  • Intended for laboratory research use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris-Acetate-EDTA
Name Note 50X Solution
CAS 77-86-1, 60-00-4, 6850-28-8
Physical Form Liquid
Quantity 1 L
DNase DNase free
Filtered Through Filtered through a 0.2-micron filter.
Purity Grade Notes DNase-, RNase- and Protease-Free
Packaging Nalgene Poly Bottle
Protease Protease free
Synonym TAE
Grade Electrophoresis
Concentration or Composition (by Analyte or Components) Tris (24%), Acetic Acid (5.0%), and EDTA (<2%).
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.