Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris Base (White Crystals or Crystalline Powder/Molecular Biology), Fisher BioReagents™

Catalog No. BP1521 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
10 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP152-1 1 kg Poly Bottle
BP152-500 500 g Poly Bottle
BP152-5 5 kg Poly Pail
BP152-10 10 kg Poly Pail
BP152-25 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. BP152-1
  • $398.50
    $261.65 / Each

    Save $136.85

    34% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,903.00
    $1,263.65 / Case of 6 EA

    Save $639.35

    34% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
This item is not returnable. View return policy

DNase-, RNase- and Protease-Free. | CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol

DNase-, RNase- and Protease-Free.

Tris Base is a buffer component in molecular biology, tissue culture, and electrophoresis procedures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-86-1
Molecular Formula C4H11NO3
Molecular Weight (g/mol) 121.136
InChI Key LENZDBCJOHFCAS-UHFFFAOYSA-N
Synonym Trimethylol Aminomethane, Tris(hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol
PubChem CID 6503
ChEBI CHEBI:9754
IUPAC Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES C(C(CO)(CO)N)O

Specifications

Absorbance 0.05 AU max. at 280nm
Melting Point 168.5°C
Boiling Point 219°C
Color White
pH 10.4
Physical Form Crystalline Powder or Crystals
Quantity 1 kg
DNase DNase free
Formulation 2-Amino-2-(Hydroxymethyl)-1,3-Propanediol, Tris (hydroxymethyl) Aminomethane, Tris Base, THAM, Trimethylol Aminomethane, Trometamol
Identification Pass Test
Assay Percent Range ≥99.8 %
Linear Formula H2NC(CH2OH)3
Packaging Poly Bottle
Protease Protease free
Merck Index 15, 9951
Formula Weight 121.14
Percent Purity ≥99.8%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Tris Base
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.