Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyl Sulfoxide, Fisher BioReagents™

Catalog No. BP231100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP231-100 100 mL Amber Glass Bottle
BP231-1 1 L Amber Glass Bottle
BP231-4 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP231-100
Get Up to $1,800 in Chemicals with Purchase
View all promotions
 Restricted Item
Add to Cart

Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 % | CAS: 67-68-5 | C2H6OS | 78.13 g/mol

Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 %

DMSO is a cryopreservation agent for cells, and may also be used in bacterial transformations.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
IUPAC Name methylsulfinylmethane
SMILES CS(C)=O

Specifications

Melting Point 18.4°C
Absorbance ≤0.30 Absorbance Units at 280nm
Color Colorless
Decomposition Information >190°C
Density 1.100g/cm³
Quantity 100 mL
Identification Pass Test
Packaging Amber Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range ≥99.7 %
Vapor Pressure 0.55mbar at 20°C
Linear Formula (CH3)2SO
Viscosity 1.98 mPa/s at 25°C
Water 0.2% max.
Merck Index 15, 3285
Boiling Point 189°C
Formula Weight 78.13
Percent Purity ≥99.7%
Chemical Name or Material Dimethyl Sulfoxide
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:2
Instability:0
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.