Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology, Fisher BioReagents™
GSA_VA

Catalog No. BP24711 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP24711 1 L Nalgene Poly Bottle
BP2471-100 100 mL Poly Bottle
BP2471500 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP24711
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

TBS, Tris Buffered Saline, 10X Solution, pH 7.4, Molecular Biology

  • Save time and simplify your buffer preparation step by using concentrated Fisher BioReagents 10X TBS buffer solution - simply dilute as needed
  • 10X solution contains 1.37M NaCl, 0.027M KCl and 0.25M tris
  • To prepare a 1X solution, mix one volume of Fisher BioReagents 10X TBS solution with 9 volumes of ultrapure water, such as BP2819
  • Filtered and autoclaved - ready to use stock TBS buffer
  • Tested for the absence of DNase, RNase, and protease
  • TBS buffer is often used in immunodetection techniques such as Western blotting protocols
  • Intended for laboratory research use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tris Buffered Saline
Name Note 10X Solution
CAS 7447-40-7, 7647-01-0, 7647-14-5, 77-86-1, 7732-18-5
Boiling Point 100°C
Color Colorless
pH 7.5
Physical Form Liquid
Quantity 1 L
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Packaging Nalgene Poly Bottle
Protease Protease free
Synonym TBS
Grade Molecular Biology
Concentration or Composition (by Analyte or Components) 10X solution contains 1.37M Sodium Chloride, 0.027M Potassium Chloride, and 0.25M Tris/Tris-HCl.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.