Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Doxycycline Hydrochloride (Yellow Powder), Fisher BioReagents™

Catalog No. BP26531 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP26531 1 g Amber Glass
BP26535 5 g Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BP26531
Get Up to $1,800 in Chemicals with Purchase
View all promotions
 Restricted Item
Add to Cart
This item is not returnable. View return policy

Broad spectrum antibiotic

Broad spectrum antibiotic. Broad spectrum inhibitor of matrix metalloproteinases in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10592-13-9
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.90
MDL Number MFCD03427564
InChI Key VLUQVUWDECWBTL-UQVCFKGQSA-N
PubChem CID 54706018
IUPAC Name [(1S,4aR,11R,11aR,12S,12aR)-3-carbamoyl-4,4a,6,7,12-pentahydroxy-11-methyl-2,5-dioxo-11,11a,12,12a-tetrahydro-1H-tetracen-1-yl]-dimethylazanium;chloride
SMILES [Cl-].C[C@@H]1[C@H]2[C@H](O)[C@H]3[C@H]([NH+](C)C)C(=O)\C(=C(/N)O)C(=O)[C@@]3(O)C(=O)C2=C(O)C2=C(O)C=CC=C12

Specifications

Type Doxycycline Hydrochloride
Physical Form Powder/Solid
Absorbance Inclusive between 300 at 349nm
Melting Point 210°C
Packaging Amber Glass
Potency Inclusive between 800μg/mg
Specific Rotation Inclusive between 105°
Water Inclusive between 1.4%
Color Yellow
Quantity 1 g
Assay Percent Range 95%
Formula Weight 480.9
Chemical Name or Material Doxycycline Hydrochloride
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not induce vomiting. Call a physician or Poison Control Center immediately. . .

NFPA

Health:2
Flammability:0
Instability:0
Recommended Storage 0° to 5°C, light sensitive, desiccate.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.