Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPES Buffer, 1M Solution, pH 7.3 (Molecular Biology), Fisher BioReagents™

Catalog No. BP299500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP299500 500 mL Poly Bottle
BP299100 100 mL Poly Bottle
BP299-1 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP299500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Commonly used buffering agent

HEPES is commonly used as a buffering agent.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HEPES Buffer
Absorbance ≤0.01(0.1M solution) at 260nm, 0.004 max. (0.1M solution) at 280nm
Name Note 1M Solution, pH 7.3
CAS 7365-45-9, 7732-18-5
Color Undesignated
Concentration 0.95 to 1.05 M
pH 7.2 to 7.5
Physical Form Liquid
Quantity 500 mL
DNase DNase free
Purity Grade Notes DNase-, RNase- and Protease-Free
Identification Pass Test
Molecular Formula C8H18N2O4S
Packaging Poly Bottle
Protease Protease free
Merck Index 15, 4689
Synonym N-(2-Hydroxyethyl)piperazine-N«-2-ethanesulfonic Acid
Formula Weight 238.197
Grade Molecular Biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.