Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Phosphate Dibasic Anhydrous (White Granules or Powder), Fisher BioReagents™

Catalog No. BP332500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP332-500 500 g Glass Bottle
BP332-1 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP332-500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Sodium Phosphate Dibasic Anhydrous (White Granules or Powder)

This buffer component is commonly used in tissue culture and molecular biology applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7558-79-4
Molecular Formula HNa2O4P
Molecular Weight (g/mol) 141.96
MDL Number MFCD00003496
InChI Key BNIILDVGGAEEIG-UHFFFAOYSA-L
Synonym disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
PubChem CID 24203
ChEBI CHEBI:34683
SMILES [Na+].[Na+].OP([O-])([O-])=O

Specifications

Color White
pH 8.7 to 9.3
Physical Form Powder/Solid
Quantity 500 g
Assay Percent Range ≥99.0 %
Merck Index 15, 8789
IUPAC Name disodium;hydrogen phosphate
Formula Weight 141.96
Percent Purity ≥99.0%
Identification Pass Test
Loss on Drying 0.2% max.
Packaging Glass Bottle
Decomposition Information Decomposes information: > 240°C
Chemical Name or Material Sodium Phosphate Dibasic Anhydrous
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Signal Word: CAUTION!
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation . The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health:1 Flammability:0 Instability:1
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.