Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Monobasic (White Crystals), Fisher BioReagents

Catalog No. BP362500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP362-500 500 g Glass Bottle
BP362-1 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP362-500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This potassium salt is used in buffers for molecular biology and cell culture applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253°C
Color White
pH 4.5
Physical Form Solid
Quantity 500 g
Assay Percent Range ≥99.0 %
Merck Index 15, 7780
IUPAC Name potassium;dihydrogen phosphate
Formula Weight 136.09
Percent Purity ≥99.0%
Identification Pass Test
Loss on Drying 0.2% max.
Packaging Glass Bottle
Decomposition Information >250°C
Chemical Name or Material Potassium Phosphate Monobasic
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.