Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Water, (For RNA Work) (DEPC-Treated, DNASE, RNASE free/Mol. Biol.), Fisher BioReagents™

Catalog No. BP5611 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Nalgene Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP561-1 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP561-1
  • $122.70
    $67.65 / Each

    Save $55.05

    45% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $580.00
    $326.65 / Case of 6 EA

    Save $253.35

    44% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

DEPC-treated and autoclaved | CAS: 7732-18-5 | H2O | 18.015 g/mol

DEPC-treated and autoclaved

Diethylpyrocarbonate (DEPC)-treated Water is guaranteed nuclease-free (tested for both DNase and RNase) and is suitable for all RNA work.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Water
pH 7
Name Note For RNA Work, DEPC-treated and DNASE, RNASE free.
Color Colorless
CAS 7732-18-5
Melting Point 0°C
Boiling Point 100°C
Molecular Formula H2O
DNase DNase free
Merck Index 15, 10237
Synonym dihydrogen oxide, dihydrogen monoxide
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name oxidane
Molecular Weight (g/mol) 18.015
PubChem CID 962
ChEBI CHEBI:15377
Formula Weight 18.02
Grade Molecular Biology
Packaging Nalgene Poly Bottle
Vapor Pressure 17.5mmHg at 20°C
Filtered Through Filtered through a 0.2-micron filter.
Treatment(s) DEPC-Treated
Density 1.000g/cm³
Quantity 1 L
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.