Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kanamycin Sulfate (White Powder), Fisher BioReagents

Catalog No. BP906-5 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP906-5 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP906-5
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

For selecting transformed cells containing kanamycin resistance gene | CAS: 25389-94-0 | C18H38N4O15S | 582.575 g/mol

For selecting transformed cells containing kanamycin resistance gene

TRUSTED_SUSTAINABILITY

Specifications

Type Kanamycin Sulfate
Chemical Name or Material Kanamycin Sulfate
Physical Form Solid
CAS 25389-94-0
pH 6.5 to 8.5
Ignition Residue < 1.0%
Packaging Poly Bottle
Potency > 750μg/mg
Color White
Quantity 5 g
Molecular Formula C18H38N4O15S
Merck Index 15, 5329
InChI Key OOYGSFOGFJDDHP-IZQIRFRQSA-N
SMILES C1C(C(C(C(C1N)OC2C(C(C(C(O2)CN)O)O)O)O)OC3C(C(C(C(O3)CO)O)N)O)N.OS(=O)(=O)O
IUPAC Name (2R,3S,4S,5S,6R)-2-(aminomethyl)-6-[(1R,2S,3S,4R,6S)-4,6-diamino-3-[(2S,3R,4S,5R,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-2-hydroxycyclohexyl]oxyoxane-3,4,5-triol;sulfuric acid
Molecular Weight (g/mol) 582.575
PubChem CID 134129479
Formula Weight 582.58
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.