Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Matrigel™ Matrix
GSA_VAGREENER_CHOICE

Catalog No. CB-40234 Shop All Corning Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
Without Additives:
No LDEV
No LDEV, No Phenol Red
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Without Additives
CB-40234 10 mL No LDEV
CB-40234A 5 mL No LDEV
CB-40234C 10 mL No LDEV, No Phenol Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer
Catalog No. CB-40234 Supplier Corning™ Supplier No. 354234
Add to Cart
Edge
Add to Cart
Note: Avoid multiple freeze-thaws; do not store in a frost-free freezer

Produces a biologically active matrix material resembling the mammalian cellular basement matrix. Corning™ Matrigel™ Matrix provides a physiologically relevant environment for studies of cell morphology, biochemical function, migration, invasion, and gene expression.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Basement Membrane Matrix
Without Additives No LDEV
For Use With (Application) Cell growth and differentiation; metabolism/toxicology studies; invasion assays; in vitro and in vivo angiogenesis assays; in vivo angiogenesis studies and augmentation of tumors in immunosuppressed mice
Product Line Matrigel
Quantity 10 mL
Source Engelbreth-Holm-Swarm Mouse Tumor
Content And Storage -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.