Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Magnesium Sulfate Anhydrous (Powder/Certified), Fisher Chemical™
GSA_VA

Catalog No. M65-500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
3 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M65-500 500 g Poly Bottle
M65-100LB 100 lb. Fiber Drum
M65-3 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. M65-500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7487-88-9
Molecular Formula MgO4S
Molecular Weight (g/mol) 120.36
MDL Number MFCD00011110
InChI Key CSNNHWWHGAXBCP-UHFFFAOYSA-L
Synonym magnesium sulfate, magnesium sulphate, magnesium sulfate anhydrous, sulfuric acid magnesium salt 1:1, bitter salt, sal angalis, magnesium sulfate 1:1, mgso4, sal de sedlitz, tomix ot
PubChem CID 24083
ChEBI CHEBI:32599
IUPAC Name magnesium(2+) sulfate
SMILES [Mg++].[O-]S([O-])(=O)=O

Specifications

Melting Point 1124°C
Color White
pH 7.9
Physical Form Solid
Quantity 500 g
Formula Weight 120.37
Grade Certified
Identification Pass Test
Packaging Poly Bottle
Decomposition Information >700°C
Density 2.65g/cm³
Chemical Name or Material Magnesium Sulfate Anhydrous
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.