Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Dibasic Anhydrous (Crystalline Powder/USP), Fisher Chemical™

Catalog No. P290500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
P290-500 500 g Poly Bottle
P290-212 2.5 kg Poly Bottle
P290-12 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P290-500
Only null left
Add to Cart

Chemical Identifiers

CAS 11-4-7758
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
MDL Number MFCD00011383
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym dipotassium hydrogen phosphate, dipotassium phosphate, dipotassium hydrogenphosphate, dibasic potassium phosphate, potassium hydrogen phosphate, potassium phosphate, dibasic, potassium dibasic phosphate, potassium phosphate dibasic, phosphoric acid, dipotassium salt, dipotassium monophosphate
PubChem CID 24450
ChEBI CHEBI:32031
IUPAC Name dipotassium;hydrogen phosphate
SMILES OP(=O)([O-])[O-].[K+].[K+]

Specifications

Melting Point 340°C
Color White
pH 8.5 to 9.6
Physical Form Solid
Quantity 500 g
Formula Weight 174.18
Percent Purity 98.0 to 100.5% (USP)
Grade USP
Identification Pass Test (USP)
Loss on Drying 1.0% max. (USP)
Packaging Poly Bottle
Decomposition Information >180°C
Chemical Name or Material Potassium Phosphate Dibasic Anhydrous
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.