Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Steriflip™ Sterile Disposable Vacuum Filter Units

Catalog No. SCGP00525 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.22 μm
0.45 μm
Material (Membrane):
PES
PVDF
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Material (Membrane)
SCGP00525 0.22 μm PES
SE1M179M6 0.22 μm PVDF
SE1M003M00 0.45 μm PVDF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SCGP00525 Supplier MilliporeSigma™ Supplier No. SCGP00525
Get 1 Extra Pack of Filtration Units With Purchase
View all promotions
Add to Cart
Add to Cart

Performance and convenience all in one. Steriflip™ Sterile Disposable Vacuum Filter Units uses vacuum to collect filtrate.

Overview
The Steriflip filter unit is designed to work with standard 50 mL centrifuge tubes, which eliminates sample transfer. To filter, just attach the unit to a 50 mL centrifuge tube containing your sample, flip it over, and apply the vacuum. When finished, the filtrate collects in the attached 50 mL centrifuge tube.

New pore sizes ideal for Cell Isolation applications

Steriflip Nylon Net filters are now available in 40, 60 and 100 μm pore sizes, making them ideal for stem cell, cardiomyocite, neurological and skeletal tissue applications. The vacuum-assisted, closed system option ensures sample sterility while enabling faster separation of large volumes of cellular material and improved recoveries.

The Steriflip™ filter unit eliminates messy pouring during sample transfer. Directly attach the unit to a 50 mL centrifuge tube containing your sample, flip it over, and apply vacuum. The filtrate collects in the attached 50 mL centrifuge tube. Steriflip™ filters come in different pore sizes that are ideal for cell isolation. These include nylon net filters in 40, 60 and 100 μm pore sizes, which are ideal for stem cell, cardiomyocyte, neurological and skeletal tissue applications. The vacuum-assisted, closed system ensures sample sterility while enabling fast separation of large volumes of cellular material for efficient recoveries.

Features & Benefits

  • Eliminates messy sample transfer via a simple vacuum-assisted, closed system
  • Isolate cells from cellular debris using any of the various pore-sized nylon net filters
  • Ideal for sterilizing small batches of culture media
  • Attaches to any standard 50 mL centrifuge tube, eliminating unnecesary materials, time and risk of spillage

Applications: Sterilization of Aqueous Solutions (up to 50 mL), Sterile Filtration of Small Batches of Culture Media, Sterile Filtration of Buffers with High Value Protein or Small Molecule Components, Cell Isolation

Order Info

Filter diameter: 40mm

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Connectors Double lead thread with vacuum port
Disposable Single-use
Material MBS/Polypropylene
Material (Membrane) PES
Pore Size 0.22 μm
Sterility Sterile
Filtration Area 7 cm2
For Use With (Application) Low protein binding clarification of aqueous solutions
No. per Case 25Pack
Quantity 25 Pack
Type Steriflip-GP Filter Unit
Volume (Metric) 50 mL
Volume (Metric) Upper 250 mL
Height (English) 5.43 in.
Height (Metric) 13.8 cm
Diameter (Metric) Filter 4 cm
Diameter (Metric) Membrane 6.4 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.