Learn More
Medchemexpress LLC HY-15667 10mg Medchemexpress, P005091 CAS:882257-11-6 Purity:>98%
SDP
Catalog No. 502029154
Encompass_Preferred
Medchemexpress, HY-15667 10mg P005091 CAS:882257-11-6 P005091 is a selective and potent inhibitor of ubiquitin-specific protease 7 (USP7) with an EC50 of 4.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9154 Supplier Medchemexpress LLC Supplier No. HY1566710MG
Medchemexpress, HY-15667 10mg P005091 CAS:882257-11-6 P005091 is a selective and potent inhibitor of ubiquitin-specific protease 7 (USP7) with an EC50 of 4.2 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.