Learn More
Azzota Corp PeriScientific Smart Peristaltic pump
Catalog No. NC3579788
Encompass_Preferred
he P1 - PeriScientific smart peristaltic pump is designed to meet your fluid transfer and dispensing needs with superior precision and accuracy, easy operation, and minimum maintenance. The ultra-bright 4.3-inch LCD touchscreen control panel interactively guides you at each step of the operation to set up sophisticated dispensing procedures in just seconds. In 4 different operation modes: Continuous Dispense Mode, Volume Dispense Mode, Repeat Dispense Mode, and Schedule Dispense Mode, the human-machine interface (HMI) accepts direct process parameter input and displays a readout of rotor speed, flow rate, dispensed volume, remaining time or elapsed time data all on one screen.
Catalog No. NC3579788 Supplier Azzota Corp Supplier No. PERIP1EZA1
he P1 - PeriScientific smart peristaltic pump is designed to meet your fluid transfer and dispensing needs with superior precision and accuracy, easy operation, and minimum maintenance. The ultra-bright 4.3-inch LCD touchscreen control panel interactively guides you at each step of the operation to set up sophisticated dispensing procedures in just seconds. In 4 different operation modes: Continuous Dispense Mode, Volume Dispense Mode, Repeat Dispense Mode, and Schedule Dispense Mode, the human-machine interface (HMI) accepts direct process parameter input and displays a readout of rotor speed, flow rate, dispensed volume, remaining time or elapsed time data all on one screen.