Learn More
Lonza Walkersville P1 Primary Cell 4D-Nucleofector X Kit L
Supplier: Lonza Walkersville V4XP1012
The P1 Primary Cell 4D-Nucleofector X Kit L is one of our five 4D-Nucleofector Kits suited for transfecting primary cells in the 100 µL Nucleocuvette Vessel format when working with the 4D-Nucleofector X Unit.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionConditions are transferable between 4D-Nucleofector System, 96-well Shuttle System and 384-well Nucleofector SystemApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cellsLow throughput applicationsContent:2 x 0.675 mL P1 Primary Cell Nucleofector Solution2 x 0.15 mL Supplement 150 µg pmaxGFP Vector (1 μg/μl in 10 mM Tris pH 8.0)12 x 100 µL Nucleocuvette Vessel12 x Single use pipettes