Learn More
Montana Molecular mNeonGreen Green Fluorescent Protein BacMam
SDP
Catalog No. 502815249
Encompass_Preferred
mNeonGreen fluorescent protein packaged in Montana Molecular's Big Sky BacMam vector. Big Sky provides robust expression in mammalian cells - even CHOs - along with many other primary and iPSC derived cultures. Use this tool to test Big Sky BacMam expression in your cells of interest.
Catalog No. 50-281-5249 Supplier Montana Molecular Supplier No. P1000G005
mNeonGreen fluorescent protein packaged in Montana Molecular's Big Sky BacMam vector. Big Sky provides robust expression in mammalian cells - even CHOs - along with many other primary and iPSC derived cultures. Use this tool to test Big Sky BacMam expression in your cells of interest.