Learn More
Medchemexpress LLC HY-114428 5mg Medchemexpress, P110δ-IN-1 CAS:1595129-71-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1144285MG
Encompass_Preferred
Medchemexpress, HY-114428 5mg P110δ-IN-1 CAS:1595129-71-7 P110δ-IN-1 is a potent and selective inhibitor of P110δ extracted from patent WO 2014055647 A1, with an IC 50 of 8.4 nM [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2490