Learn More
Cayman Chemical p120CATnInCTerm RabbIt 100uL
SDP
Catalog No. 502950473
Encompass_Preferred
An MTH1 inhibitor (IC50 72 nM) selective for MTH1 over a panel of 456 kinases at 1 UM induces oxidative DNA damage and apoptosis in SGC-7901 and BGC-823 cancer cells at 30 UM induces ROS production and apoptosis in the same model inhibits proliferation in a panel of 14 cancer cell lines expressing wild-type or various mutant forms of K-Ras or p53 (IC50s 0.52-7.4 UM) inhibits tumor growth in an SW 480 mouse xenograft model at 25 mg/kg
Catalog No. 50-295-0473 Supplier Cayman Chemical Supplier No. 32283100 UL
An MTH1 inhibitor (IC50 72 nM) selective for MTH1 over a panel of 456 kinases at 1 UM induces oxidative DNA damage and apoptosis in SGC-7901 and BGC-823 cancer cells at 30 UM induces ROS production and apoptosis in the same model inhibits proliferation in a panel of 14 cancer cell lines expressing wild-type or various mutant forms of K-Ras or p53 (IC50s 0.52-7.4 UM) inhibits tumor growth in an SW 480 mouse xenograft model at 25 mg/kg