Learn More
Werther International P15TC SILENT COMPRESSOR 115V
SDP
Catalog No. 501037791
Encompass_Preferred
Silent air compressor, P15TC 115V/60Hz, Output 0.7 CFM / 20 l.min, Max Pressure 120 PSI, Noise Level 30 dB/A, 0.93 Gal air tank, 2 year warranty, Dimensions 11 in.L x 13 in.W x 14 in.H
Catalog No. 50-103-7791 Supplier Werther International Supplier No. M01090
Silent air compressor, P15TC 115V/60Hz, Output 0.7 CFM / 20 l.min, Max Pressure 120 PSI, Noise Level 30 dB/A, 0.93 Gal air tank, 2 year warranty, Dimensions 11 in.L x 13 in.W x 14 in.H