Learn More
Diagnostic Biosystems P16 p16
Catalog No. 502407509
Encompass_Preferred
P16 is a mitotic inhibitor protein. It competes with D-type cyclins to bind to cdk4 and cdk6. It acts as tumor suppressor and inhibits the progression of cells through the G1 phase of the cell cycle.
Catalog No. 50-240-7509 Supplier Diagnostic Biosystems Supplier No. PDM575
P16 is a mitotic inhibitor protein. It competes with D-type cyclins to bind to cdk4 and cdk6. It acts as tumor suppressor and inhibits the progression of cells through the G1 phase of the cell cycle.