Learn More
StatLab P16 Cervical Cancer Control Slides
SDP
Catalog No. 21033344
Change view
Click to view available options
Product Type:
Control Slide
Catalog No. Product Type
21-033-344 Control Slide
1 options
Catalog No. 21-033-344 Supplier StatLab Supplier No. CSP16/15
Add to Cart
Add to Cart
Use P16 cervical cancer control slides with confidence to evaluate stain quality and performance in histology laboratories
  • Slides consist of formalin fixed paraffin embedded tissue sections which contain known distinctive tissue elements
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Includes 15 Unstained Slides per pack
Specifications
Product Type Control Slide