Learn More
Fluotics Tecan Fluent Evo MCA compatible P200, non-sterile
Catalog No. NC1972312
Encompass
Tecan Fluent Evo MCA compatible P200, non-sterile, for MCA head, working volume: 200 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 30038616
Catalog No. NC1972312 Supplier Fluotics Supplier No. MCA200XL.NS
Tecan Fluent Evo MCA compatible P200, non-sterile, for MCA head, working volume: 200 uL, Case Size: 50 racks/case with 96 tips per rack, Compare to: 30038616