Learn More
enQuire Bioreagents p21WAF1 Anti-Human Monoclonal [Clone SPM306], Size: 100ug
Supplier:  enQuire Bioreagents 1026MSM2XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM306. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2a kappa. Positive Control: HeLa Cells. Skin, colon, or breast carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Human recombinant p21 protein. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 21kDa.Chromosome: 6p21.31. Gene Symbol: CDKN1A. Gene ID: 1026. Ensemble: ENSG00000124762. Uniprot: NP_510867.
Catalog No. 50-188-4557