Learn More
TARGETMOL CHEMICALS INC P22074 5MG
SDP
Catalog No. 502580526
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. P22074 is a USP7 inhibitor. It is not an active antagonist like its halogenated related compounds. p22074 has antitumour activity. purity: 100%
Catalog No. 50-258-0526 Supplier TARGETMOL CHEMICALS INC Supplier No. T282845MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. P22074 is a USP7 inhibitor. It is not an active antagonist like its halogenated related compounds. p22074 has antitumour activity. purity: 100%