Learn More
Enzo Life Sciences p23 polyclonal antibody (50µl)
SDP
Supplier:  Enzo Life Sciences ADISPA670D
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human p23. UniProt ID: Q15185. Source: Whole rabbit serum. Species reactivity: Human, Mouse, Rat. Applications: WB. Recommended Dilutions: Western Blot (1:1,000, colorimetric) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Antiserum. Formulation: Serum. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7452