Learn More
AUTOMATION TRAINER P250 Beckman-Type AP96, Unfiltered, Non-Sterile, Racked, Solid Base
SDP
Catalog No. NC2231852
Encompass_Preferred
Automation Trainers pipette tips are designed to be a direct replacement to Beckmans AP96 P20, P50, & P250 tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate labware definition is not required.- Fits Biomek 3000, NX, NXp, FX and FXp Laboratory Automation Workstations - High-Grade Polypropylene Build - P20 (Case of 50 96-Tip Racks), P50 (Case of 50 96-Tip Racks), and P250 (Case of 50 96-Tip Racks)
Catalog No. NC2231852 Supplier AUTOMATION TRAINER Supplier No. BP250AP96UNRB
Automation Trainers pipette tips are designed to be a direct replacement to Beckmans AP96 P20, P50, & P250 tip. Our tips are manufactured to strict standards to ensure quality and consistent performance. Automation control software protocol modification and separate labware definition is not required.- Fits Biomek 3000, NX, NXp, FX and FXp Laboratory Automation Workstations - High-Grade Polypropylene Build - P20 (Case of 50 96-Tip Racks), P50 (Case of 50 96-Tip Racks), and P250 (Case of 50 96-Tip Racks)