Learn More
enQuire Bioreagents p27Kip1 Anti-Human Monoclonal [Clone SPM348], Size: 100ug
Supplier:  enQuire Bioreagents 1027MSM1XP1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: SPM348. Clonality: Monoclonal. Host: Mouse. Isotype: IgG1 kappa. Positive Control: ZR75, T47D, SK-BR-3, MDA-MB-231, MCF7 cells. Tonsil, Breast or Colon Ca. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Purified GST-p27 fusion protein of mouse origin. Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 25-26kDa.Chromosome: 12p13.1. Gene Symbol: CDKN1B. Gene ID: 1027. Ensemble: ENSG00000111276. Uniprot: NP_004055.
Catalog No. 50-188-4562