Learn More
enQuire Bioreagents p27Kip1 Anti-Human Monoclonal [Clone DCS-72.F6 + KIP1/769], Size: 100ug
Supplier:  enQuire Bioreagents 1027MSM4P1
Encompass_Preferred
Product Type: Antibody. Application: Flow, IF, WB, IHC. Conjugate/Tag: Unconjugated. Clone: DCS-72.F6 + KIP1/769. Clonality: Monoclonal. Host: Mouse. Isotype: IgG. Positive Control: ZR75, T47D, SK-BR-3, MDA-MB-231, HeLa or MCF7 cells. Tonsil, Breast, Cervical or Colon Carcinoma. Cellular Staining: Nuclear. Buffer: 10mM PBS with 0.05% BSA & 0.05% azide. Concentration: 200ug/ml. Purity: Protein A/G Purified. Immunogen: Mouse recombinant p27 protein (DCS-72.F6); Recombinant human CDKN1B protein (KIP1/769). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 25-26kDa.Chromosome: 12p13.1. Gene Symbol: CDKN1B. Gene ID: 1027. Ensemble: ENSG00000111276. Uniprot: NP_004055.
Catalog No. 50-188-4565