Learn More
Cayman Chemical p27KIp1CTerm RabbIt Monc 100uL
SDP
Catalog No. 502948600
Encompass_Preferred
A BET family protein inhibitor inhibits binding of BRD2 BRD3 and BRD4 to an acetylated histone H4 peptide in vitro decreases Myc levels in and inhibits proliferation of AML DLBCL and MM cells reduces tumor growth in AML DLBCL and MM mouse xenograft models reduces tumor growth when administered alone or in combination with docetaxel or MDV 3100 in a 22Rv1 CRPC mouse xenograft model
Catalog No. 50-294-8600 Supplier Cayman Chemical Supplier No. 32247100 UL
A BET family protein inhibitor inhibits binding of BRD2 BRD3 and BRD4 to an acetylated histone H4 peptide in vitro decreases Myc levels in and inhibits proliferation of AML DLBCL and MM cells reduces tumor growth in AML DLBCL and MM mouse xenograft models reduces tumor growth when administered alone or in combination with docetaxel or MDV 3100 in a 22Rv1 CRPC mouse xenograft model